The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022087.3 | NP_071370.2 | 1 - 2 | 212 | 62 |
XM_017012499.1 | XP_016867988.1 | 1 - 2 | 193 | 62 | |
XM_017012500.1 | XP_016867989.1 | 1 - 2 | 193 | 62 | |
GenBank mRNA | DA308896.1 | - | 1 - 2 | 135 | 62 |
DA748534.1 | - | 1 - 2 | 193 | 62 |