Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOTCH4
Entrez Gene ID
4855
Gene Name
notch 4
Gene Aliases
INT3
Chromosome Location
Chr.6: 32194843 - 32224067 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004557.3 NP_004548.3 10 - 11 1871 108
NR_134949.1 11 - 12 1973 108
NR_134950.1 10 - 11 1871 108
GenBank mRNA AK290785.1 - 10 - 11 1868 108
BC063815.1 - 10 - 11 1861 108
BC140782.1 - 10 - 11 1826 108
BC144540.1 - 11 - 12 1934 108
BC144541.1 - 10 - 11 1826 108
U95299.1 - 10 - 11 1822 108

View Full Product Details