Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOTCH4
Entrez Gene ID
4855
Gene Name
notch 4
Gene Aliases
INT3
Chromosome Location
Chr.6: 32194843 - 32224067 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004557.3 NP_004548.3 24 - 25 4675 102
NR_134949.1 24 - 25 4253 102
NR_134950.1 23 - 24 4151 102
GenBank mRNA AK131314.1 - 3 - 4 3014 102
BC140782.1 - 24 - 25 4630 102
BC144540.1 - 24 - 25 4214 102
D63395.1 - 7 - 8 1812 102
U95299.1 - 24 - 25 4626 102

View Full Product Details