Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B4GALNT2
Entrez Gene ID
124872
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 2
Gene Aliases
B4GALT, GALGT2
Chromosome Location
Chr.17: 49120347 - 49170722 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159387.1 NP_001152859.1 1 - 2 38 60

View Full Product Details