The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130849.1 | NP_001124321.1 | 4 - 5 | 953 | 136 |
NM_001130850.1 | NP_001124322.1 | 4 - 5 | 697 | 136 | |
NM_016289.3 | NP_057373.1 | 4 - 5 | 831 | 136 | |
XM_011511350.2 | XP_011509652.1 | 4 - 5 | 831 | 136 | |
GenBank mRNA | AF113536.1 | - | 3 - 4 | 452 | 136 |
AF134480.1 | - | 3 - 4 | 404 | 136 | |
AF151824.1 | - | 3 - 4 | 399 | 136 | |
AK000804.1 | - | 3 - 4 | 334 | 136 | |
AK292382.1 | - | 4 - 5 | 953 | 136 | |
AK315692.1 | - | 4 - 5 | 665 | 136 | |
BC020570.1 | - | 4 - 5 | 723 | 136 |