The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135776.1 | NP_001129248.1 | 2 - 2 | 4984 | 68 |
NM_014007.3 | NP_054726.1 | 3 - 3 | 5107 | 68 | |
XM_005251835.3 | XP_005251892.1 | 3 - 3 | 5428 | 68 | |
XM_011518409.1 | XP_011516711.1 | 4 - 4 | 5120 | 68 | |
XM_011518411.2 | XP_011516713.1 | 2 - 2 | 5234 | 68 |