The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080850.2 | NP_001074319.1 | 1 - 2 | 98 | 159 |
XM_011542067.2 | XP_011540369.1 | 2 - 3 | 1030 | 159 | |
XM_011542068.2 | XP_011540370.1 | 2 - 3 | 1035 | 159 | |
XM_011542069.2 | XP_011540371.1 | 2 - 3 | 1027 | 159 | |
XM_011542071.2 | XP_011540373.1 | 2 - 3 | 1023 | 159 | |
XM_017002225.1 | XP_016857714.1 | 1 - 2 | 115 | 159 | |
XM_017002226.1 | XP_016857715.1 | 2 - 3 | 426 | 159 | |
GenBank mRNA | AK024625.1 | - | 5 - 6 | 533 | 159 |
AK302075.1 | - | 4 - 5 | 420 | 159 | |
DC397297.1 | - | 4 - 5 | 422 | 159 |