The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018067.1 | NP_001018077.1 | 4 - 5 | 836 | 128 |
NM_001018068.1 | NP_001018078.1 | 4 - 5 | 818 | 128 | |
GenBank mRNA | AK074970.1 | - | 4 - 5 | 785 | 128 |
BC002488.2 | - | 4 - 5 | 745 | 128 | |
BC003049.1 | - | 4 - 5 | 744 | 128 | |
BC008045.2 | - | 4 - 5 | 767 | 128 | |
BC019273.1 | - | 4 - 5 | 773 | 128 | |
BC020555.1 | - | 4 - 5 | 750 | 128 | |
BC026916.1 | - | 4 - 5 | 768 | 128 |