The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003680.3 | NP_003671.1 | 10 - 11 | 1930 | 126 |
XM_011542347.1 | XP_011540649.1 | 8 - 9 | 929 | 126 | |
XM_011542348.1 | XP_011540650.1 | 9 - 10 | 997 | 126 | |
XM_017002651.1 | XP_016858140.1 | 11 - 12 | 1303 | 126 | |
GenBank mRNA | AK026535.1 | - | 11 - 12 | 1223 | 126 |
AK125213.1 | - | 10 - 11 | 2051 | 126 | |
AK127182.1 | - | 2 - 3 | 1452 | 126 | |
AK223608.1 | - | 10 - 11 | 1256 | 126 | |
BC001933.1 | - | 10 - 11 | 1164 | 126 | |
BC004151.2 | - | 10 - 11 | 1164 | 126 | |
BC016689.1 | - | 10 - 11 | 1189 | 126 | |
BC035242.1 | - | 1 - 2 | 1059 | 126 | |
KU178431.1 | - | 10 - 11 | 1142 | 126 | |
U89436.1 | - | 10 - 11 | 1142 | 126 |