Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PKHD1
Entrez Gene ID
5314
Gene Name
polycystic kidney and hepatic disease 1 (autosomal recessive)
Gene Aliases
ARPKD, FCYT, TIGM1
Chromosome Location
Chr.6: 51613291 - 52087625 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138694.3 NP_619639.3 44 - 45 7382 79
NM_170724.2 NP_733842.2 44 - 45 7382 79
XM_011514680.2 XP_011512982.1 44 - 45 7726 79
XM_011514682.2 XP_011512984.1 44 - 45 7348 79
XM_011514683.2 XP_011512985.1 45 - 46 6706 79
XM_011514684.2 XP_011512986.1 36 - 37 6492 79
XM_011514685.1 XP_011512987.1 44 - 45 7382 79
XM_011514686.2 XP_011512988.1 44 - 45 7382 79
XM_011514687.1 XP_011512989.1 44 - 45 7382 79
XM_011514688.2 XP_011512990.1 44 - 45 7348 79
XM_011514690.2 XP_011512992.1 9 - 10 1268 79
XM_011514691.2 XP_011512993.1 9 - 10 7293 79
XM_017010944.1 XP_016866433.1 44 - 45 7208 79
XM_017010945.1 XP_016866434.1 43 - 44 7273 79
XM_017010946.1 XP_016866435.1 44 - 45 7348 79
XM_017010947.1 XP_016866436.1 42 - 43 7084 79
XM_017010948.1 XP_016866437.1 35 - 36 7166 79
XM_017010949.1 XP_016866438.1 26 - 27 6345 79
XM_017010950.1 XP_016866439.1 44 - 45 7382 79
XM_017010951.1 XP_016866440.1 44 - 45 7348 79
XM_017010952.1 XP_016866441.1 44 - 45 7348 79
GenBank mRNA AF480064.1 - 44 - 45 7328 79
AK128031.1 - 8 - 9 1305 79
AY074797.1 - 44 - 45 7382 79
AY092083.1 - 44 - 45 7368 79

View Full Product Details