Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PKHD1
Entrez Gene ID
5314
Gene Name
polycystic kidney and hepatic disease 1 (autosomal recessive)
Gene Aliases
ARPKD, FCYT, TIGM1
Chromosome Location
Chr.6: 51613291 - 52087625 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138694.3 NP_619639.3 49 - 50 8186 69
NM_170724.2 NP_733842.2 49 - 50 8186 69
XM_011514680.2 XP_011512982.1 49 - 50 8530 69
XM_011514682.2 XP_011512984.1 49 - 50 8152 69
XM_011514683.2 XP_011512985.1 50 - 51 7510 69
XM_011514684.2 XP_011512986.1 41 - 42 7296 69
XM_011514685.1 XP_011512987.1 49 - 50 8186 69
XM_011514686.2 XP_011512988.1 49 - 50 8186 69
XM_011514687.1 XP_011512989.1 49 - 50 8186 69
XM_011514688.2 XP_011512990.1 49 - 50 8152 69
XM_011514690.2 XP_011512992.1 14 - 15 2072 69
XM_011514691.2 XP_011512993.1 14 - 15 8097 69
XM_017010944.1 XP_016866433.1 49 - 50 8012 69
XM_017010945.1 XP_016866434.1 48 - 49 8077 69
XM_017010946.1 XP_016866435.1 49 - 50 8152 69
XM_017010947.1 XP_016866436.1 47 - 48 7888 69
XM_017010948.1 XP_016866437.1 40 - 41 7970 69
XM_017010949.1 XP_016866438.1 31 - 32 7149 69
XM_017010950.1 XP_016866439.1 49 - 50 8186 69
XM_017010951.1 XP_016866440.1 49 - 50 8152 69
XM_017010952.1 XP_016866441.1 49 - 50 8152 69
GenBank mRNA AF480064.1 - 49 - 50 8132 69
AK128031.1 - 13 - 14 2109 69
AY074797.1 - 49 - 50 8186 69
AY092083.1 - 49 - 50 8172 69

View Full Product Details