Product Details

Assay Reference Genome
Location

Chr.11:113410568 on build GRCh38
Cytoband
11q23.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
1813
Gene Symbol
DRD2
Gene Name
dopamine receptor D2
Gene Aliases
D2DR, D2R
Location
Chr.11:113409595-113475279 on build GRCh38
Assay Gene Location
Within Exon 9
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
DRD2 NM_000795.3 8 1726 NP_000786.1
NM_016574.3 7 1639 NP_057658.2
XM_017017296.1 8 1708 XP_016872785.1
AB209832.1 7 1521 BAD93069.1
AF176812.1 AAF61479.1
BC021195.2 8 1586 AAH21195.1
M29066.1 7 1524 AAA52761.1
S62137.1 7 1808 AAB26819.1
S69899.1 7 1503 AAB20571.1
X51362.1 8 1656 CAA35746.1
X51645.1 CAB56463.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1040152 Chr11:113387343 - 113415194 on Build GRCh38 Loss ANKK1 DRD2
nsv556444 Chr11:113318201 - 113512672 on Build GRCh38 Gain ANKK1 TTC12 DRD2 MIR4301

View Full Product Details