The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242377.1 | NP_001229306.1 | 4 - 5 | 654 | 164 |
NM_152624.5 | NP_689837.2 | 4 - 5 | 654 | 164 | |
NR_038352.1 | 2 - 3 | 374 | 164 | ||
XM_017009163.1 | XP_016864652.1 | 5 - 6 | 746 | 164 | |
GenBank mRNA | AK090564.1 | - | 4 - 5 | 569 | 164 |
AK296361.1 | - | 2 - 3 | 304 | 164 | |
AY135173.1 | - | 4 - 5 | 628 | 164 | |
AY146650.1 | - | 4 - 5 | 628 | 164 | |
BC064593.1 | - | 4 - 5 | 560 | 164 | |
BG720562.1 | - | 2 - 3 | 379 | 165 |