The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301065.1 | NP_001287994.1 | 12 - 13 | 1498 | 93 |
NM_007180.2 | NP_009111.2 | 13 - 14 | 1591 | 93 | |
GenBank mRNA | AB000824.1 | - | 13 - 14 | 1601 | 93 |
AK091981.1 | - | 11 - 12 | 1460 | 93 | |
AK223140.1 | - | 13 - 14 | 1565 | 93 | |
AK223143.1 | - | 13 - 14 | 1569 | 93 | |
BC109206.2 | - | 12 - 13 | 1492 | 93 |