Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHRF1
Entrez Gene ID
57661
Gene Name
PHD and ring finger domains 1
Gene Aliases
PPP1R125, RNF221
Chromosome Location
Chr.11: 576446 - 612225 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286581.1 NP_001273510.1 13 - 14 1774 89
NM_001286582.1 NP_001273511.1 13 - 14 1768 89
NM_001286583.1 NP_001273512.1 13 - 14 1762 89
NM_020901.3 NP_065952.2 13 - 14 1771 89
XM_005253025.4 XP_005253082.1 13 - 14 1732 89
XM_005253027.3 XP_005253084.1 13 - 14 1720 89
XM_011520236.2 XP_011518538.1 13 - 14 1768 89
XM_011520237.2 XP_011518539.1 12 - 13 1615 89
GenBank mRNA AB040975.1 - 13 - 14 1734 89
BC112931.1 - 11 - 12 3516 89
BC136615.1 - 13 - 14 1699 89
BC144293.1 - 13 - 14 1696 89
BC144294.1 - 13 - 14 1687 89
BC144295.1 - 13 - 14 1693 89
BC146771.1 - 13 - 14 1734 89

View Full Product Details