The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286581.1 | NP_001273510.1 | 15 - 16 | 4582 | 85 |
NM_001286582.1 | NP_001273511.1 | 15 - 16 | 4576 | 85 | |
NM_001286583.1 | NP_001273512.1 | 15 - 16 | 4570 | 85 | |
NM_020901.3 | NP_065952.2 | 15 - 16 | 4579 | 85 | |
XM_005253025.4 | XP_005253082.1 | 15 - 16 | 4540 | 85 | |
XM_005253027.3 | XP_005253084.1 | 15 - 16 | 4528 | 85 | |
XM_011520236.2 | XP_011518538.1 | 15 - 16 | 4576 | 85 | |
XM_011520237.2 | XP_011518539.1 | 14 - 15 | 4423 | 85 | |
GenBank mRNA | AB040975.1 | - | 15 - 16 | 4542 | 85 |
BC004950.1 | - | 2 - 3 | 434 | 85 | |
BC013381.2 | - | 2 - 3 | 1058 | 85 | |
BC029651.1 | - | 2 - 3 | 2708 | 85 | |
BC112931.1 | - | 13 - 14 | 6312 | 85 | |
BC136615.1 | - | 15 - 16 | 4507 | 85 | |
BC144293.1 | - | 15 - 16 | 4504 | 85 | |
BC144294.1 | - | 15 - 16 | 4495 | 85 | |
BC144295.1 | - | 15 - 16 | 4501 | 85 | |
BC146771.1 | - | 15 - 16 | 4542 | 85 |