Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHRF1
Entrez Gene ID
57661
Gene Name
PHD and ring finger domains 1
Gene Aliases
PPP1R125, RNF221
Chromosome Location
Chr.11: 576446 - 612225 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286581.1 NP_001273510.1 5 - 6 673 109
NM_001286582.1 NP_001273511.1 5 - 6 670 109
NM_001286583.1 NP_001273512.1 5 - 6 661 109
NM_020901.3 NP_065952.2 5 - 6 673 109
XM_005253025.4 XP_005253082.1 5 - 6 631 109
XM_005253027.3 XP_005253084.1 5 - 6 619 109
XM_011520236.2 XP_011518538.1 5 - 6 673 109
XM_011520237.2 XP_011518539.1 4 - 5 514 109
GenBank mRNA BC136615.1 - 5 - 6 598 109
BC144293.1 - 5 - 6 598 109
BC144294.1 - 5 - 6 586 109
BC144295.1 - 5 - 6 595 109

View Full Product Details