Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAMK2G
Entrez Gene ID
818
Gene Name
calcium/calmodulin dependent protein kinase II gamma
Gene Aliases
CAMK, CAMK-II, CAMKG
Chromosome Location
Chr.10: 73812501 - 73874591 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320898.1 NP_001307827.1 17 - 18 1314 69
NM_172171.2 NP_751911.1 16 - 17 1287 69
XM_005270195.1 XP_005270252.1 17 - 18 1341 69
XM_005270197.1 XP_005270254.1 17 - 18 1305 69
XM_005270198.1 XP_005270255.1 17 - 18 1299 69
XM_005270199.1 XP_005270256.1 16 - 17 1272 69
XM_005270200.1 XP_005270257.1 16 - 17 1242 69
XM_005270201.1 XP_005270258.1 16 - 17 1236 69
XM_006717993.1 XP_006718056.1 18 - 19 1374 69
XM_006717996.1 XP_006718059.1 17 - 18 1311 69
XM_011540225.2 XP_011538527.1 16 - 17 1226 69
XM_017016716.1 XP_016872205.1 18 - 19 1374 69
XM_017016717.1 XP_016872206.1 18 - 19 1322 69
XM_017016718.1 XP_016872207.1 17 - 18 1341 69
XM_017016719.1 XP_016872208.1 18 - 19 1332 69
XM_017016720.1 XP_016872209.1 17 - 18 1311 69
XM_017016721.1 XP_016872210.1 18 - 19 1449 69
XM_017016722.1 XP_016872211.1 17 - 18 1299 69
XM_017016723.1 XP_016872212.1 16 - 17 1278 69
XM_017016724.1 XP_016872213.1 16 - 17 1272 69
XM_017016725.1 XP_016872214.1 16 - 17 1242 69
XM_017016726.1 XP_016872215.1 16 - 17 1236 69
XM_017016727.1 XP_016872216.1 15 - 16 1209 69
XM_017016728.1 XP_016872217.1 16 - 17 1226 69
XM_017016729.1 XP_016872218.1 16 - 17 1109 69
XM_017016731.1 XP_016872220.1 16 - 17 1217 69
XM_017016732.1 XP_016872221.1 16 - 17 1147 69
XM_017016734.1 XP_016872223.1 16 - 17 1237 69
XM_017016737.1 XP_016872226.1 15 - 16 1180 69
XM_017016738.1 XP_016872227.1 17 - 18 1240 69
XM_017016739.1 XP_016872228.1 18 - 19 1506 69
GenBank mRNA AK095576.1 - 16 - 17 1326 69
U32472.1 - 3 - 4 140 69
U32473.1 - 3 - 4 182 69
U32509.1 - 2 - 3 113 69
U66063.1 - 6 - 7 332 69

View Full Product Details