The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042734.3 | NP_001036199.1 | 9 - 10 | 2065 | 95 |
NM_001300813.2 | NP_001287742.1 | 11 - 12 | 2260 | 95 | |
NM_001318085.1 | NP_001305014.1 | 10 - 11 | 2167 | 95 | |
NM_001318086.1 | NP_001305015.1 | 9 - 10 | 2062 | 95 | |
NM_006323.4 | NP_006314.2 | 10 - 11 | 2170 | 95 | |
XM_005262688.1 | XP_005262745.1 | 11 - 12 | 2139 | 95 | |
XM_005262691.1 | XP_005262748.1 | 10 - 11 | 2031 | 95 | |
XM_011531535.2 | XP_011529837.1 | 10 - 11 | 2034 | 95 | |
XM_011531536.2 | XP_011529838.1 | 10 - 11 | 2243 | 95 | |
XM_011531537.2 | XP_011529839.1 | 10 - 11 | 1949 | 95 | |
XM_011531539.2 | XP_011529841.1 | 9 - 10 | 1851 | 95 | |
XM_011531540.2 | XP_011529842.1 | 5 - 6 | 680 | 95 | |
XM_017007652.1 | XP_016863141.1 | 8 - 9 | 1784 | 95 | |
GenBank mRNA | AJ131245.1 | - | 10 - 11 | 2170 | 95 |
AK300278.1 | - | 9 - 10 | 1853 | 95 | |
AK304234.1 | - | 7 - 8 | 910 | 95 | |
BC040137.1 | - | 10 - 11 | 2028 | 95 | |
BC117135.1 | - | 9 - 10 | 2008 | 95 | |
BC143268.1 | - | 11 - 12 | 2203 | 95 | |
BC143276.1 | - | 9 - 10 | 2008 | 95 |