The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040402.2 | NP_001035492.1 | 5 - 6 | 586 | 66 |
NM_001287755.1 | NP_001274684.1 | 5 - 6 | 482 | 66 | |
NM_001287757.1 | NP_001274686.1 | 6 - 7 | 749 | 66 | |
NM_015115.3 | NP_055930.2 | 5 - 6 | 586 | 66 | |
XM_005265731.3 | XP_005265788.1 | 5 - 6 | 476 | 66 | |
XM_011534379.2 | XP_011532681.1 | 5 - 6 | 1009 | 66 | |
XM_011534380.1 | XP_011532682.1 | 5 - 6 | 345 | 66 | |
XM_017007912.1 | XP_016863401.1 | 5 - 6 | 439 | 66 | |
XM_017007913.1 | XP_016863402.1 | 4 - 5 | 318 | 66 | |
GenBank mRNA | AK027051.1 | - | 5 - 6 | 405 | 66 |
AK124346.1 | - | 4 - 5 | 546 | 66 | |
AK294894.1 | - | 5 - 6 | 482 | 66 | |
AK294896.1 | - | 6 - 7 | 596 | 66 | |
BC053897.1 | - | 5 - 6 | 452 | 66 | |
BX537944.1 | - | 5 - 6 | 413 | 66 | |
D87466.1 | - | 5 - 6 | 396 | 66 | |
DA770685.1 | - | 5 - 6 | 476 | 66 |