Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCUN1D4
Entrez Gene ID
23142
Gene Name
defective in cullin neddylation 1 domain containing 4
Gene Aliases
-
Chromosome Location
Chr.4: 51843000 - 51916837 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040402.2 NP_001035492.1 7 - 8 744 156
NM_001287755.1 NP_001274684.1 7 - 8 640 156
NM_001287757.1 NP_001274686.1 8 - 9 907 156
NM_015115.3 NP_055930.2 7 - 8 744 156
XM_005265731.3 XP_005265788.1 7 - 8 634 156
XM_011534379.2 XP_011532681.1 7 - 8 1167 156
XM_011534380.1 XP_011532682.1 7 - 8 503 156
XM_017007912.1 XP_016863401.1 7 - 8 597 156
XM_017007913.1 XP_016863402.1 6 - 7 1082 156
GenBank mRNA AK027051.1 - 7 - 8 563 156
AK294894.1 - 7 - 8 640 156
AK294896.1 - 8 - 9 754 156
BC053897.1 - 7 - 8 610 156
BX537944.1 - 7 - 8 571 156
D87466.1 - 7 - 8 554 156

View Full Product Details