Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCUN1D4
Entrez Gene ID
23142
Gene Name
defective in cullin neddylation 1 domain containing 4
Gene Aliases
-
Chromosome Location
Chr.4: 51843000 - 51916837 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040402.2 NP_001035492.1 9 - 10 964 112
NM_001287755.1 NP_001274684.1 9 - 10 860 112
NM_001287757.1 NP_001274686.1 10 - 11 1127 112
XM_011534379.2 XP_011532681.1 9 - 10 1387 112
XM_011534380.1 XP_011532682.1 9 - 10 723 112
XM_017007913.1 XP_016863402.1 8 - 9 1302 112
GenBank mRNA AK027051.1 - 9 - 10 783 112
AK294894.1 - 9 - 10 860 112
AK294896.1 - 10 - 11 974 112
BX537944.1 - 9 - 10 791 112
D87466.1 - 9 - 10 774 112

View Full Product Details