The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272011.1 | NP_001258940.1 | 9 - 10 | 1125 | 65 |
NM_014226.2 | NP_055041.1 | 10 - 11 | 1215 | 65 | |
XM_011537053.2 | XP_011535355.1 | 4 - 5 | 1242 | 65 | |
XM_017021549.1 | XP_016877038.1 | 10 - 11 | 1212 | 65 | |
XM_017021550.1 | XP_016877039.1 | 9 - 10 | 1111 | 65 | |
XM_017021551.1 | XP_016877040.1 | 11 - 12 | 1155 | 65 | |
XM_017021552.1 | XP_016877041.1 | 10 - 11 | 1253 | 65 | |
XM_017021553.1 | XP_016877042.1 | 8 - 9 | 1050 | 65 | |
XM_017021554.1 | XP_016877043.1 | 8 - 9 | 1049 | 65 | |
XM_017021559.1 | XP_016877048.1 | 9 - 10 | 1204 | 65 | |
GenBank mRNA | AB022694.1 | - | 10 - 11 | 1209 | 65 |
AK302349.1 | - | 9 - 10 | 1110 | 65 | |
AK312778.1 | - | 10 - 11 | 1206 | 65 | |
BC026069.1 | - | 9 - 10 | 1626 | 65 | |
BC043179.1 | - | 8 - 9 | 1971 | 65 | |
BC053536.1 | - | 9 - 10 | 2170 | 65 |