The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024829.5 | NP_079105.4 | 8 - 9 | 1342 | 146 |
GenBank mRNA | AK026315.1 | - | 8 - 9 | 1105 | 146 |
AK225458.1 | - | 8 - 9 | 1105 | 146 | |
AK290914.1 | - | 8 - 9 | 1203 | 146 | |
BC000909.2 | - | 2 - 3 | 305 | 146 | |
BC063561.1 | - | 8 - 9 | 1206 | 146 |