Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTPS2
Entrez Gene ID
56474
Gene Name
CTP synthase 2
Gene Aliases
-
Chromosome Location
Chr.X: 16587999 - 16712979 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144002.1 NP_001137474.1 10 - 11 1837 91
NM_019857.4 NP_062831.3 10 - 11 1390 91
NM_175859.2 NP_787055.1 10 - 11 1476 91
XM_005274562.3 XP_005274619.1 10 - 11 1253 91
XM_006724503.3 XP_006724566.1 11 - 12 1428 91
XM_011545545.2 XP_011543847.1 10 - 11 1249 91
GenBank mRNA AF226667.1 - 10 - 11 1293 91
AK023549.1 - 10 - 11 1348 91
AK024070.1 - 10 - 11 1331 91
AK125332.1 - 10 - 11 1837 91
AK125348.1 - 10 - 11 1467 91
BC006256.2 - 8 - 9 832 91
BC034986.1 - 10 - 11 1455 91

View Full Product Details