The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144002.1 | NP_001137474.1 | 17 - 18 | 2434 | 73 |
NM_019857.4 | NP_062831.3 | 17 - 18 | 1987 | 73 | |
NM_175859.2 | NP_787055.1 | 17 - 18 | 2073 | 73 | |
XM_005274562.3 | XP_005274619.1 | 17 - 18 | 1850 | 73 | |
XM_006724503.3 | XP_006724566.1 | 18 - 19 | 2025 | 73 | |
XM_011545545.2 | XP_011543847.1 | 17 - 18 | 1846 | 73 | |
GenBank mRNA | AF226667.1 | - | 17 - 18 | 1890 | 73 |
AK023549.1 | - | 17 - 18 | 1945 | 73 | |
AK024070.1 | - | 17 - 18 | 1928 | 73 | |
AK125332.1 | - | 17 - 18 | 2434 | 73 | |
AK125348.1 | - | 17 - 18 | 2064 | 73 | |
AK309508.1 | - | 2 - 3 | 490 | 73 | |
BC006256.2 | - | 15 - 16 | 1429 | 73 | |
BC034986.1 | - | 17 - 18 | 2052 | 73 |