Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLRC1
Entrez Gene ID
3821
Gene Name
killer cell lectin like receptor C1
Gene Aliases
CD159A, NKG2, NKG2A
Chromosome Location
Chr.12: 10442264 - 10454685 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007328.3 NP_015567.1 3 - 4 492 59
NM_213657.2 NP_998822.1 4 - 5 666 59
GenBank mRNA BC012550.1 - 4 - 5 670 59
X54868.1 - 3 - 4 442 59

View Full Product Details