Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBAP2L
Entrez Gene ID
9898
Gene Name
ubiquitin associated protein 2 like
Gene Aliases
NICE-4, NICE4
Chromosome Location
Chr.1: 154220172 - 154271510 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127320.2 NP_001120792.1 2 - 3 205 70
NM_001287815.1 NP_001274744.1 2 - 3 259 70
NM_001287816.1 NP_001274745.1 2 - 3 259 70
NM_014847.3 NP_055662.3 2 - 3 259 70
XM_005245658.3 XP_005245715.1 2 - 3 259 70
XM_005245667.3 XP_005245724.1 2 - 3 259 70
XM_005245668.3 XP_005245725.1 2 - 3 259 70
XM_005245669.3 XP_005245726.1 2 - 3 259 70
XM_005245670.3 XP_005245727.1 2 - 3 259 70
XM_005245672.3 XP_005245729.1 2 - 3 184 70
XM_005245673.3 XP_005245730.1 2 - 3 259 70
XM_005245674.1 XP_005245731.1 2 - 3 184 70
XM_011510215.2 XP_011508517.1 2 - 3 259 70
XM_011510216.1 XP_011508518.1 2 - 3 184 70
XM_017002972.1 XP_016858461.1 2 - 3 284 70
XM_017002973.1 XP_016858462.1 2 - 3 184 70
XM_017002974.1 XP_016858463.1 2 - 3 215 70
XM_017002975.1 XP_016858464.1 2 - 3 259 70
XM_017002976.1 XP_016858465.1 2 - 3 161 70
XM_017002977.1 XP_016858466.1 2 - 3 184 70
XM_017002978.1 XP_016858467.1 2 - 3 215 70
XM_017002979.1 XP_016858468.1 2 - 3 176 70
XM_017002980.1 XP_016858469.1 2 - 3 215 70
XM_017002981.1 XP_016858470.1 2 - 3 176 70
XM_017002982.1 XP_016858471.1 2 - 3 184 70
XM_017002983.1 XP_016858472.1 2 - 3 215 70
XM_017002984.1 XP_016858473.1 2 - 3 215 70
XM_017002985.1 XP_016858474.1 2 - 3 284 70
XM_017002986.1 XP_016858475.1 2 - 3 184 70
XM_017002987.1 XP_016858476.1 2 - 3 215 70
XM_017002988.1 XP_016858477.1 2 - 3 215 70
XM_017002989.1 XP_016858478.1 2 - 3 215 70
XM_017002990.1 XP_016858479.1 2 - 3 176 70
XM_017002991.1 XP_016858480.1 1 - 2 117 70
GenBank mRNA AJ243668.1 - 2 - 3 203 70
AK124294.1 - 2 - 3 191 70
AK302662.1 - 2 - 3 193 70
AK303533.1 - 2 - 3 186 70
BC003170.1 - 1 - 2 141 70
D63478.1 - 2 - 3 198 70
DC369803.1 - 2 - 3 205 70

View Full Product Details