Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABTB2
Entrez Gene ID
25841
Gene Name
ankyrin repeat and BTB domain containing 2
Gene Aliases
ABTB2A, BTBD22
Chromosome Location
Chr.11: 34150987 - 34358008 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145804.2 NP_665803.2 15 - 16 3189 82
GenBank mRNA AK056863.1 - 3 - 4 245 82
AK091560.1 - 12 - 13 1407 82
AK095632.1 - 15 - 16 2436 82
AK291744.1 - 15 - 16 3190 82
BC093968.1 - 15 - 16 2301 82
BX640841.1 - 7 - 8 890 82

View Full Product Details