The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145804.2 | NP_665803.2 | 8 - 9 | 2280 | 87 |
GenBank mRNA | AK091560.1 | - | 5 - 6 | 498 | 87 |
AK095632.1 | - | 8 - 9 | 1527 | 87 | |
AK291744.1 | - | 8 - 9 | 2281 | 87 | |
BC093968.1 | - | 8 - 9 | 1392 | 87 |