Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EPN1
Entrez Gene ID
29924
Gene Name
epsin 1
Gene Aliases
-
Chromosome Location
Chr.19: 55675195 - 55695767 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130071.1 NP_001123543.1 10 - 11 2331 110
NM_001130072.1 NP_001123544.1 10 - 11 1868 110
NM_013333.3 NP_037465.2 9 - 10 1790 110
XM_005258829.2 XP_005258886.1 10 - 11 1877 110
XM_005258830.2 XP_005258887.1 10 - 11 1865 110
XM_011526880.1 XP_011525182.1 10 - 11 1874 110
XM_011526881.1 XP_011525183.1 9 - 10 1802 110
XM_017026722.1 XP_016882211.1 9 - 10 1799 110
XM_017026723.1 XP_016882212.1 12 - 13 1919 110
XM_017026724.1 XP_016882213.1 12 - 13 1857 110
XM_017026725.1 XP_016882214.1 11 - 12 1844 110
GenBank mRNA AF073727.1 - 10 - 11 2330 110
AK022454.1 - 11 - 12 1837 110
AK300545.1 - 11 - 12 1738 110
AK314690.1 - 10 - 11 1835 110
BC044651.1 - 9 - 10 1740 110

View Full Product Details