The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130071.1 | NP_001123543.1 | 7 - 8 | 1870 | 60 |
NM_001130072.1 | NP_001123544.1 | 7 - 8 | 1407 | 60 | |
NM_013333.3 | NP_037465.2 | 6 - 7 | 1332 | 60 | |
XM_005258829.2 | XP_005258886.1 | 7 - 8 | 1416 | 60 | |
XM_005258830.2 | XP_005258887.1 | 7 - 8 | 1407 | 60 | |
XM_011526880.1 | XP_011525182.1 | 7 - 8 | 1416 | 60 | |
XM_011526881.1 | XP_011525183.1 | 6 - 7 | 1341 | 60 | |
XM_017026722.1 | XP_016882211.1 | 6 - 7 | 1341 | 60 | |
XM_017026723.1 | XP_016882212.1 | 9 - 10 | 1458 | 60 | |
XM_017026724.1 | XP_016882213.1 | 9 - 10 | 1396 | 60 | |
XM_017026725.1 | XP_016882214.1 | 8 - 9 | 1383 | 60 | |
GenBank mRNA | AF073727.1 | - | 7 - 8 | 1869 | 60 |
AK022454.1 | - | 7 - 8 | 1376 | 60 | |
AK300545.1 | - | 8 - 9 | 1280 | 60 | |
AK314690.1 | - | 7 - 8 | 1377 | 60 | |
BC044651.1 | - | 6 - 7 | 1282 | 60 |