Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
U2AF2
Entrez Gene ID
11338
Gene Name
U2 small nuclear RNA auxiliary factor 2
Gene Aliases
U2AF65
Chromosome Location
Chr.19: 55654050 - 55674716 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012478.1 NP_001012496.1 4 - 5 1390 62
NM_007279.2 NP_009210.1 4 - 5 1390 62
XM_006722994.2 XP_006723057.1 5 - 6 1428 62
XM_011526410.1 XP_011524712.1 5 - 6 1428 62
GenBank mRNA AB451261.1 - 4 - 5 335 62
AB451387.1 - 4 - 5 335 62
BC008740.2 - 4 - 5 394 62
BC030574.2 - 4 - 5 1294 62
X64044.1 - 4 - 5 375 62

View Full Product Details