The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020366.3 | NP_065099.3 | 19 - 20 | 3240 | 76 |
GenBank mRNA | AF227257.1 | - | 8 - 9 | 1668 | 76 |
AF260257.1 | - | 10 - 11 | 2192 | 76 | |
AF265666.1 | - | 8 - 9 | 1667 | 76 | |
AF265667.1 | - | 8 - 9 | 1433 | 76 | |
AJ417067.1 | - | 19 - 20 | 3240 | 76 | |
AK301780.1 | - | 7 - 8 | 1298 | 76 | |
BC039089.1 | - | 7 - 8 | 1283 | 76 |