Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPGRIP1
Entrez Gene ID
57096
Gene Name
retinitis pigmentosa GTPase regulator interacting protein 1
Gene Aliases
CORD13, LCA6, RGI1, RGRIP, RPGRIP, RPGRIP1d
Chromosome Location
Chr.14: 21287977 - 21351316 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020366.3 NP_065099.3 12 - 13 1613 98
XM_005267879.2 XP_005267936.1 3 - 4 605 98
XM_005267880.2 XP_005267937.1 3 - 4 605 98
XM_011536978.1 XP_011535280.1 3 - 4 605 98
XM_011536979.1 XP_011535281.1 3 - 4 605 98
XM_011536980.1 XP_011535282.1 3 - 4 605 98
XM_011536981.1 XP_011535283.1 3 - 4 605 98
XM_011536982.1 XP_011535284.1 3 - 4 605 98
XM_017021473.1 XP_016876962.1 3 - 4 605 98
GenBank mRNA AF260257.1 - 3 - 4 565 98
AJ417067.1 - 12 - 13 1613 98
AK301780.1 - 3 - 4 619 98
AY914561.1 - 1 - 2 32 98
BC039089.1 - 3 - 4 604 98

View Full Product Details