The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020366.3 | NP_065099.3 | 4 - 5 | 582 | 119 |
GenBank mRNA | AJ417067.1 | - | 4 - 5 | 582 | 119 |