Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPGRIP1
Entrez Gene ID
57096
Gene Name
retinitis pigmentosa GTPase regulator interacting protein 1
Gene Aliases
CORD13, LCA6, RGI1, RGRIP, RPGRIP, RPGRIP1d
Chromosome Location
Chr.14: 21287977 - 21351316 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020366.3 NP_065099.3 7 - 8 928 63
GenBank mRNA AJ417067.1 - 7 - 8 928 63

View Full Product Details