The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288614.1 | NP_001275543.1 | 3 - 4 | 630 | 67 |
GenBank mRNA | BX647523.1 | - | 3 - 4 | 328 | 67 |