The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001076780.1 | NP_001070248.1 | 13 - 14 | 2256 | 106 |
NM_001278423.1 | NP_001265352.1 | 2 - 3 | 350 | 106 | |
NM_001278425.1 | NP_001265354.1 | 2 - 3 | 350 | 106 | |
NM_052892.3 | NP_443124.3 | 13 - 14 | 2256 | 106 | |
NR_126532.1 | 13 - 14 | 2256 | 106 | ||
GenBank mRNA | AB067466.1 | - | 5 - 6 | 601 | 106 |
AK098052.1 | - | 7 - 8 | 838 | 106 | |
AY164483.1 | - | 13 - 14 | 2256 | 106 | |
AY371495.1 | - | 2 - 3 | 312 | 106 | |
BC004562.2 | - | 2 - 3 | 317 | 106 | |
BC014157.2 | - | 2 - 3 | 317 | 106 | |
DA326413.1 | - | 2 - 3 | 319 | 106 |