Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PKD1L2
Entrez Gene ID
114780
Gene Name
polycystin 1 like 2 (gene/pseudogene)
Gene Aliases
PC1L2
Chromosome Location
Chr.16: 81100879 - 81220370 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076780.1 NP_001070248.1 17 - 18 2911 70
NM_001278423.1 NP_001265352.1 6 - 7 1005 70
GenBank mRNA AK098052.1 - 11 - 12 1493 70
BC004562.2 - 6 - 7 972 70
BC014157.2 - 6 - 7 972 70

View Full Product Details