The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142930.1 | NP_001136402.1 | 4 - 5 | 561 | 144 |
NM_001142931.1 | NP_001136403.1 | 3 - 4 | 399 | 144 | |
NM_006595.3 | NP_006586.1 | 4 - 5 | 561 | 144 | |
NR_024625.1 | 5 - 6 | 623 | 144 | ||
XM_006718359.3 | XP_006718422.1 | 4 - 5 | 551 | 144 | |
XM_017018464.1 | XP_016873953.1 | 5 - 6 | 608 | 144 | |
XM_017018465.1 | XP_016873954.1 | 5 - 6 | 608 | 144 | |
GenBank mRNA | AF229253.1 | - | 3 - 4 | 175 | 144 |
AF229254.1 | - | 4 - 5 | 501 | 144 | |
AK294724.1 | - | 3 - 4 | 356 | 144 | |
AK299288.1 | - | 4 - 5 | 527 | 144 | |
AK304157.1 | - | 5 - 6 | 589 | 144 | |
AY311389.1 | - | 4 - 5 | 500 | 144 | |
BC017709.1 | - | 4 - 5 | 481 | 144 | |
BT007093.1 | - | 4 - 5 | 388 | 144 | |
U83857.1 | - | 4 - 5 | 483 | 144 | |
Y15906.1 | - | 4 - 5 | 520 | 144 |