Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LPIN2
Entrez Gene ID
9663
Gene Name
lipin 2
Gene Aliases
-
Chromosome Location
Chr.18: 2916994 - 3013315 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014646.2 NP_055461.1 15 - 16 2325 91
XM_005258177.4 XP_005258234.1 15 - 16 2395 91
XM_005258178.3 XP_005258235.1 15 - 16 2324 91
XM_005258179.4 XP_005258236.1 15 - 16 2258 91
XM_017026098.1 XP_016881587.1 16 - 17 2414 91
XM_017026099.1 XP_016881588.1 15 - 16 2185 91
GenBank mRNA BC152448.1 - 15 - 16 2325 91
D87436.1 - 15 - 16 2325 91

View Full Product Details