The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301016.2 | NP_001287945.1 | 9 - 10 | 1390 | 72 |
NM_001321208.1 | NP_001308137.1 | 9 - 10 | 1430 | 72 | |
NM_001321211.1 | NP_001308140.1 | 9 - 10 | 1214 | 72 | |
NM_007040.5 | NP_008971.2 | 9 - 10 | 1721 | 72 | |
NM_144732.4 | NP_653333.1 | 9 - 10 | 1203 | 72 | |
XM_005258459.3 | XP_005258516.1 | 9 - 10 | 1524 | 72 | |
XM_005258461.4 | XP_005258518.1 | 9 - 10 | 1354 | 72 | |
XM_005258463.3 | XP_005258520.1 | 9 - 10 | 1345 | 72 | |
XM_005258464.3 | XP_005258521.1 | 9 - 10 | 1160 | 72 | |
XM_005258465.4 | XP_005258522.1 | 9 - 10 | 1371 | 72 | |
XM_011526392.2 | XP_011524694.1 | 9 - 10 | 1524 | 72 | |
XM_011526393.2 | XP_011524695.1 | 9 - 10 | 1446 | 72 | |
XM_011526394.2 | XP_011524696.1 | 9 - 10 | 1201 | 72 | |
XM_011526395.2 | XP_011524697.1 | 9 - 10 | 1410 | 72 | |
GenBank mRNA | AJ007509.1 | - | 9 - 10 | 1564 | 72 |
AK022863.1 | - | 9 - 10 | 1523 | 72 | |
AK023006.1 | - | 9 - 10 | 1160 | 72 | |
AK127057.1 | - | 8 - 9 | 1181 | 72 | |
AK225850.1 | - | 9 - 10 | 1183 | 72 | |
AK290729.1 | - | 9 - 10 | 1185 | 72 | |
AK291315.1 | - | 10 - 11 | 1470 | 72 | |
AK291762.1 | - | 9 - 10 | 1523 | 72 | |
AK297127.1 | - | 9 - 10 | 1318 | 72 | |
AK308156.1 | - | 10 - 11 | 1203 | 72 | |
AL050146.1 | - | 2 - 3 | 329 | 72 | |
BC002564.2 | - | 9 - 10 | 1461 | 72 | |
BC009988.2 | - | 9 - 10 | 1367 | 72 | |
BC014232.2 | - | 9 - 10 | 1342 | 72 | |
BC027713.2 | - | 9 - 10 | 1444 | 72 |