The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297549.1 | NP_001284478.1 | 17 - 18 | 2764 | 83 |
NM_002911.3 | NP_002902.2 | 17 - 18 | 2731 | 83 | |
XM_017027105.1 | XP_016882594.1 | 17 - 18 | 2797 | 83 | |
XM_017027106.1 | XP_016882595.1 | 17 - 18 | 2764 | 83 | |
GenBank mRNA | AF074016.1 | - | 17 - 18 | 2683 | 83 |
AK128727.1 | - | 2 - 3 | 1213 | 83 | |
BC039817.1 | - | 17 - 18 | 2728 | 83 | |
D86988.1 | - | 17 - 18 | 2698 | 83 | |
U65533.1 | - | 17 - 18 | 2687 | 83 |