The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003104.5 | NP_003095.2 | 5 - 6 | 723 | 71 |
NR_034039.1 | 5 - 6 | 808 | 71 | ||
GenBank mRNA | AK295656.1 | - | 4 - 5 | 486 | 71 |
AK296574.1 | - | 5 - 6 | 717 | 71 | |
AK309672.1 | - | 5 - 6 | 717 | 71 | |
AK312444.1 | - | 5 - 6 | 696 | 71 | |
BC021085.2 | - | 5 - 6 | 673 | 71 | |
BC025295.1 | - | 5 - 6 | 621 | 71 | |
FJ224316.1 | - | 5 - 6 | 673 | 71 | |
L29008.1 | - | 5 - 6 | 680 | 71 | |
U07361.1 | - | 5 - 6 | 684 | 71 |