Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AKR1D1
Entrez Gene ID
6718
Gene Name
aldo-keto reductase family 1, member D1
Gene Aliases
3o5bred, CBAS2, SRD5B1
Chromosome Location
Chr.7: 138076432 - 138118304 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK058142.1 - 6 - 7 604 126

View Full Product Details