The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014502.4 | NP_055317.1 | 7 - 8 | 769 | 60 |
GenBank mRNA | AJ131186.1 | - | 7 - 8 | 769 | 60 |
BC008719.2 | - | 7 - 8 | 764 | 60 | |
BC018665.2 | - | 7 - 8 | 764 | 60 | |
BC018698.2 | - | 7 - 8 | 764 | 60 |