The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004392.6 | NP_004383.4 | 4 - 5 | 1689 | 69 |
NM_080759.5 | NP_542937.3 | 8 - 9 | 2295 | 69 | |
NM_080760.5 | NP_542938.3 | 5 - 6 | 1851 | 69 | |
XM_011534939.2 | XP_011533241.1 | 9 - 10 | 3327 | 69 | |
XM_011534942.2 | XP_011533244.1 | 7 - 8 | 1161 | 69 | |
XM_017020396.1 | XP_016875885.1 | 8 - 9 | 1321 | 69 | |
GenBank mRNA | AF102546.1 | - | 8 - 9 | 2295 | 69 |
AF356492.1 | - | 8 - 9 | 2295 | 69 | |
AJ005670.1 | - | 8 - 9 | 1548 | 69 | |
AK001000.1 | - | 9 - 10 | 1186 | 69 | |
AL079278.1 | - | 8 - 9 | 1552 | 69 | |
BC021219.2 | - | 8 - 9 | 1317 | 69 |