The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000815.4 | NP_000806.2 | 2 - 3 | 278 | 59 |
XM_011541194.2 | XP_011539496.1 | 2 - 3 | 2190 | 59 | |
XM_017000936.1 | XP_016856425.1 | 1 - 2 | 3324 | 59 | |
GenBank mRNA | AF016917.1 | - | 2 - 3 | 183 | 59 |
AK290861.1 | - | 2 - 3 | 259 | 59 | |
BC033801.1 | - | 2 - 3 | 194 | 59 | |
BI824761.1 | - | 2 - 3 | 202 | 59 | |
DA060315.1 | - | 2 - 3 | 278 | 59 | |
DA064930.1 | - | 2 - 3 | 209 | 59 |