Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UNC79
Entrez Gene ID
57578
Gene Name
unc-79 homolog (C. elegans)
Gene Aliases
KIAA1409
Chromosome Location
Chr.14: 93333219 - 93710473 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020818.3 NP_065869.3 13 - 14 1652 72
XM_011537018.2 XP_011535320.1 13 - 14 4069 72
XM_011537020.2 XP_011535322.1 13 - 14 4069 72
XM_011537021.2 XP_011535323.1 13 - 14 1620 72
XM_011537022.2 XP_011535324.1 13 - 14 4069 72
XM_011537023.2 XP_011535325.1 13 - 14 3910 72
XM_011537024.2 XP_011535326.1 13 - 14 4069 72
XM_011537025.2 XP_011535327.1 13 - 14 4069 72
XM_011537026.2 XP_011535328.1 13 - 14 4562 72
XM_011537027.2 XP_011535329.1 13 - 14 1587 72
XM_011537028.2 XP_011535330.1 13 - 14 1652 72
XM_011537029.2 XP_011535331.1 4 - 5 535 72
XM_017021506.1 XP_016876995.1 13 - 14 4069 72
XM_017021507.1 XP_016876996.1 13 - 14 4069 72
XM_017021508.1 XP_016876997.1 13 - 14 4069 72
XM_017021509.1 XP_016876998.1 10 - 11 3910 72
XM_017021510.1 XP_016876999.1 13 - 14 4069 72
XM_017021511.1 XP_016877000.1 8 - 9 863 72
XM_017021512.1 XP_016877001.1 13 - 14 1620 72
XM_017021513.1 XP_016877002.1 13 - 14 4069 72
XM_017021514.1 XP_016877003.1 13 - 14 4069 72
XM_017021515.1 XP_016877004.1 13 - 14 1468 72
XM_017021516.1 XP_016877005.1 13 - 14 4069 72
XM_017021517.1 XP_016877006.1 13 - 14 3910 72
XM_017021518.1 XP_016877007.1 7 - 8 1991 72
XM_017021519.1 XP_016877008.1 3 - 4 1085 72
GenBank mRNA AB037830.2 - 10 - 11 940 72
AK125327.1 - 13 - 14 1652 72

View Full Product Details